Stearyl stearate, 98%, Thermo Scientific Chemicals
Stearyl stearate, 98%, Thermo Scientific Chemicals
Stearyl stearate, 98%, Thermo Scientific Chemicals
Stearyl stearate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Stearyl stearate, 98%, Thermo Scientific Chemicals
CAS: 2778-96-3 | C36H72O2 | 536.97 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number FSA368110010
Price (MYR)
937.00
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
937.00
Add to cart
Chemical Identifiers
CAS626-02-8
IUPAC Name3-iodophenol
Molecular FormulaC6H5IO
InChI KeyFXTKWBZFNQHAAO-UHFFFAOYSA-N
SMILESOC1=CC=CC(I)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)or fused solid
Melting point37°C to 43°C
Appearance (Color)White to yellow to brown or grey or purple
Appearance (Form)Crystalline powder or crystals and/or chunks