Pyridine, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™, Thermo Scientific Chemicals
Pyridine, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™, Thermo Scientific Chemicals
Pyridine, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™, Thermo Scientific Chemicals
Pyridine, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Pyridine, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™, Thermo Scientific Chemicals
Pyridine, 99.5%, C5H5N, CAS Number-110-86-1, azabenzene, azine, caswell no. 717, piridina, piridina italian, pirydyna, pirydyna polish, pyridin, pyridin german, rcra waste number u196, 1L, Vapor Pressure: 20mbar at 20 deg.C, 20, 181, 115.0 deg.C to 116.0 deg.C, CHEBI:16227, Colorless, 0.9780g/mL
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
2.5 L
Packaging:
AcroSeal™ Glass Bottle
Catalog number FSA364425000
Price (MYR)
733.00
Add to cart
Quantity:
500 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (MYR)
733.00
Add to cart
Chemical Identifiers
CAS86891-03-4
IUPAC Namesodium 4-oxopent-2-en-2-olate
Molecular FormulaC5H7NaO2
InChI KeyAIWZOHBYSFSQGV-UHFFFAOYSA-M
SMILES[Na+].CC([O-])=CC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
CommentMay contain small amounts of sodium hydroxide
Assay (Non-aqueous acid-base Titration)≥92.0 to ≤108.0% (dry weight basis)
Water Content (Karl Fischer Titration)≤26.95%