Isopropanol, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™, Thermo Scientific Chemicals
Isopropanol, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™, Thermo Scientific Chemicals
Isopropanol, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™, Thermo Scientific Chemicals
Isopropanol, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™, Thermo Scientific Chemicals
Isopropanol, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Isopropanol, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
Packaging:
View all
Glass Bottle
Catalog number FSA364405000
Price (MYR)
415.00
Add to cart
Quantity:
500 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
415.00
Add to cart
Chemical Identifiers
CAS821-48-7
IUPAC Namebis(2-chloroethyl)azanium chloride
Molecular FormulaC4H10Cl3N
InChI KeyYMDZDFSUDFLGMX-UHFFFAOYSA-N
SMILES[Cl-].ClCC[NH2+]CCCl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale brown
FormCrystals or powder or crystalline powder or flakes
Assay (Titration ex Chloride)≥97.5 to ≤102.5%
Identification (FTIR)Conforms
Melting Point (clear melt)212.0-220.0?C