4-Thiouracil, 97%
4-Thiouracil, 97%
4-Thiouracil, 97%
Thermo Scientific Chemicals
4-Thiouracil, 97%
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Packaging:
Glass bottle
Catalog number FSA359930010
Price (MYR)
937.00
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
937.00
Add to cart
Chemical Identifiers
CAS91-21-4
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.5655-1.5710 @ 20°C
Appearance (Color)Clear colorless to yellow
FormLiquid or viscous liquid
Assay (GC)≥95.0%