Check Order Status
3-chloro-2-methylbenzenesulfonyl chloride, 98%, Thermo Scientific™
Element Search
Structure Search
Thermo Scientific Chemicals
3-chloro-2-methylbenzenesulfonyl chloride, 98%, Thermo Scientific™
Have Questions?
Change view
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number
FSA357080010
Price (MYR)
320.00
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
320.00
Add to cart
Specifications
Chemical Name or Material
3-chloro-2-methylbenzenesulfonyl chloride
CAS
80563-86-6
CAS Min %
97.5
Health Hazard 1
GHS Signal Word:
Danger
CAS Max %
100.0