Methyl sulfoxide, 99.8+%, for peptide synthesis, Thermo Scientific Chemicals
Methyl sulfoxide, 99.8+%, for peptide synthesis, Thermo Scientific Chemicals
Methyl sulfoxide, 99.8+%, for peptide synthesis, Thermo Scientific Chemicals
Methyl sulfoxide, 99.8+%, for peptide synthesis, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methyl sulfoxide, 99.8+%, for peptide synthesis, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
Packaging:
Glass Bottle
Catalog number FSA354845000
Price (MYR)
606.00
Add to cart
Quantity:
500 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
606.00
Add to cart
Specifications
Chemical Name or MaterialMethyl sulfoxide
Melting Point18.4°C
Name NoteFor peptide synthesis
CAS67-68-5