1-Butyl-3-methylimidazolium tetrafluoroborate, 98+%, Thermo Scientific Chemicals
1-Butyl-3-methylimidazolium tetrafluoroborate, 98+%, Thermo Scientific Chemicals
1-Butyl-3-methylimidazolium tetrafluoroborate, 98+%, Thermo Scientific Chemicals
1-Butyl-3-methylimidazolium tetrafluoroborate, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Butyl-3-methylimidazolium tetrafluoroborate, 98+%, Thermo Scientific Chemicals
CAS: 174501-65-6 | C8H15BF4N2 | 226.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Packaging:
Glass bottle
Catalog number FSA354210250
Price (MYR)
787.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
787.00
Add to cart
Chemical Identifiers
CAS148980-31-8
IUPAC Nameterbium(3+) tritrifluoromethanesulfonate
Molecular FormulaC3F9O9S3Tb
InChI KeyJLYAVYSVLLIWEK-UHFFFAOYSA-K
SMILES[Tb+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder
Assay (unspecified)Terbium: ≥25.6% (theoretical: 26.2%)
Ionic liquid
RUO – Research Use Only