1-Butyl-3-methylimidazolium chloride, 98+%, Thermo Scientific Chemicals
1-Butyl-3-methylimidazolium chloride, 98+%, Thermo Scientific Chemicals
1-Butyl-3-methylimidazolium chloride, 98+%, Thermo Scientific Chemicals
1-Butyl-3-methylimidazolium chloride, 98+%, Thermo Scientific Chemicals
1-Butyl-3-methylimidazolium chloride, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Butyl-3-methylimidazolium chloride, 98+%, Thermo Scientific Chemicals
CAS: 79917-90-1 | C8H15ClN2 | 174.67 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
500 g
Catalog number FSA354091000
Price (MYR)
1,517.00
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
1,517.00
Add to cart
Chemical Identifiers
CAS10051-44-2
IUPAC Namesodium hexanoate
Molecular FormulaC6H11NaO2
InChI KeyUDWXLZLRRVQONG-UHFFFAOYSA-M
SMILES[Na+].CCCCCC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Powder
Infrared spectrumConforms
Loss on drying=<2 %
Titration with HClO4>=98.5 %
Suitable for battery materials development.
Ionic liquid
RUO – Research Use Only