1-Butyl-3-methylimidazolium chloride, 98+%, Thermo Scientific Chemicals
1-Butyl-3-methylimidazolium chloride, 98+%, Thermo Scientific Chemicals
1-Butyl-3-methylimidazolium chloride, 98+%, Thermo Scientific Chemicals
1-Butyl-3-methylimidazolium chloride, 98+%, Thermo Scientific Chemicals
1-Butyl-3-methylimidazolium chloride, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Butyl-3-methylimidazolium chloride, 98+%, Thermo Scientific Chemicals
CAS: 79917-90-1 | C8H15ClN2 | 174.67 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
500 g
Catalog number FSA354090250
Price (MYR)
606.00
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
606.00
Add to cart
Chemical Identifiers
CAS121-34-6
IUPAC Name4-hydroxy-3-methoxybenzoic acid
Molecular FormulaC8H8O4
InChI KeyWKOLLVMJNQIZCI-UHFFFAOYSA-N
SMILESCOC1=CC(=CC=C1O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Silylated GC)≥97.5% (non-U.S. sourced material)
Appearance (Color)White to cream to yellow to pale brown
FormPowder
CommentMaterial Sourced in the U.S. and in other countries
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5% (non-U.S. sourced material)
View more
Suitable for battery materials development.
Ionic liquid
RUO – Research Use Only