1-Butyl-3-methylimidazolium chloride, 98+%, Thermo Scientific Chemicals
1-Butyl-3-methylimidazolium chloride, 98+%, Thermo Scientific Chemicals
1-Butyl-3-methylimidazolium chloride, 98+%, Thermo Scientific Chemicals
1-Butyl-3-methylimidazolium chloride, 98+%, Thermo Scientific Chemicals
1-Butyl-3-methylimidazolium chloride, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Butyl-3-methylimidazolium chloride, 98+%, Thermo Scientific Chemicals
CAS: 79917-90-1 | C8H15ClN2 | 174.67 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
500 g
Catalog number FSA354090050
Price (MYR)
464.00
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
464.00
Add to cart
Chemical Identifiers
CAS3133-81-1
IUPAC Namemethyl 3-methyl-1-benzothiophene-2-carboxylate
Molecular FormulaC11H10O2S
InChI KeyFDBXIMKCWBPTDG-UHFFFAOYSA-N
SMILESCOC(=O)C1=C(C)C2=CC=CC=C2S1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
FormCrystals or powder or crystalline powder
Melting Point (clear melt)99.5-108.5?C
Appearance (Color)White to pale cream
Suitable for battery materials development.
Ionic liquid
RUO – Research Use Only