Morin hydrate, Thermo Scientific Chemicals
Morin hydrate, Thermo Scientific Chemicals
Morin hydrate, Thermo Scientific Chemicals
Morin hydrate, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Morin hydrate, Thermo Scientific Chemicals
CAS: 654055-01-3 | C15H10O7 | 302.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
5 g
10 g
Packaging:
Glass bottle
Catalog number FSA354000500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS723-62-6
IUPAC Nameanthracene-9-carboxylic acid
Molecular FormulaC15H10O2
InChI KeyXGWFJBFNAQHLEF-UHFFFAOYSA-N
SMILESOC(=O)C1=C2C=CC=CC2=CC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to yellow-green
FormPowder
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
Clarity0.5% w/v in ethanol: clear solution
Indicator
RUO – Research Use Only