Isophorondiamine, 99+%, mixture of cis and trans, Thermo Scientific Chemicals
Isophorondiamine, 99+%, mixture of cis and trans, Thermo Scientific Chemicals
Isophorondiamine, 99+%, mixture of cis and trans, Thermo Scientific Chemicals
Isophorondiamine, 99+%, mixture of cis and trans, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Isophorondiamine, 99+%, mixture of cis and trans, Thermo Scientific Chemicals
CAS: 2855-13-2 | C10H22N2 | 170.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
250 mL
Packaging:
Glass bottle
Catalog number FSA352940010
Price (MYR)
784.00
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
784.00
Add to cart
Chemical Identifiers
CAS2687-91-4
IUPAC Name1-ethylpyrrolidin-2-one
Molecular FormulaC6H11NO
InChI KeyZFPGARUNNKGOBB-UHFFFAOYSA-N
SMILESCCN1CCCC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4640 to 1.4680 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=97.5 %
Appearance (Color)Clear colorless to yellow