L-Ascorbic acid sodium salt, 99%
L-Ascorbic acid sodium salt, 99%
L-Ascorbic acid sodium salt, 99%
L-Ascorbic acid sodium salt, 99%
Thermo Scientific Chemicals
L-Ascorbic acid sodium salt, 99%
CAS: 134-03-2 | C6H7NaO6 | 198.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
500 g
Packaging:
View all
Plastic bottle
Catalog number FSA352681000
Price (MYR)
453.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
453.00
Add to cart
Specifications
Chemical Name or MaterialL-Ascorbic acid sodium salt
Name Note99%
CAS134-03-2
Melting Point219.0°C to 221.0°C
ColorWhite to Yellow