L-Ascorbic acid sodium salt, 99%
L-Ascorbic acid sodium salt, 99%
L-Ascorbic acid sodium salt, 99%
L-Ascorbic acid sodium salt, 99%
Thermo Scientific Chemicals
L-Ascorbic acid sodium salt, 99%
CAS: 134-03-2 | C6H7NaO6 | 198.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
Packaging:
View all
Glass bottle
Catalog number FSA352680050
Price (MYR)
251.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
251.00
Add to cart
Chemical Identifiers
CAS1310-73-2
IUPAC Namesodium hydroxide
Molecular FormulaHNaO
InChI KeyHEMHJVSKTPXQMS-UHFFFAOYSA-M
SMILES[OH-].[Na+]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Strength0.995 to 1.005 N (20°C)
Appearance (Form)Solution