trans-4-Methoxycinnamaldehyde, 98%, Thermo Scientific Chemicals
trans-4-Methoxycinnamaldehyde, 98%, Thermo Scientific Chemicals
trans-4-Methoxycinnamaldehyde, 98%, Thermo Scientific Chemicals
trans-4-Methoxycinnamaldehyde, 98%, Thermo Scientific Chemicals
trans-4-Methoxycinnamaldehyde, 98%, Thermo Scientific Chemicals
CAS: 24680-50-0 | C10H10O2 | 162.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA351710250
Price (MYR)
184.42
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
184.42
Add to cart
Chemical Identifiers
CAS51-21-8
IUPAC Name5-fluoro-1,2,3,4-tetrahydropyrimidine-2,4-dione
Molecular FormulaC4H3FN2O2
InChI KeyGHASVSINZRGABV-UHFFFAOYSA-N
SMILESFC1=CNC(=O)NC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration with NaOH98.5 to 101.0 % (On dried substance)
Heavy metals (as Pb)=<20 ppm
Sulfated ash=<0.1 %
Loss on drying=<0.5 % (80°C, 4 hrs) (in vacuum)