trans-4-Methoxycinnamaldehyde, 98%, Thermo Scientific Chemicals
trans-4-Methoxycinnamaldehyde, 98%, Thermo Scientific Chemicals
trans-4-Methoxycinnamaldehyde, 98%, Thermo Scientific Chemicals
trans-4-Methoxycinnamaldehyde, 98%, Thermo Scientific Chemicals
trans-4-Methoxycinnamaldehyde, 98%, Thermo Scientific Chemicals
CAS: 24680-50-0 | C10H10O2 | 162.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number FSA351710050
Price (MYR)
67.13
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
67.13
Add to cart
Chemical Identifiers
CAS619-45-4
IUPAC Namemethyl 4-aminobenzoate
Molecular FormulaC8H9NO2
InChI KeyLZXXNPOYQCLXRS-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=C(N)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration with HClO4>=97.5 %
Appearance (Form)Crystalline powder or crystals
Appearance (Color)White to beige
Melting point110°C to 113°C