Pyridine-d5, for NMR, packaged in 0.75 ml ampoules, 99.8 atom% D
Pyridine-d5, for NMR, packaged in 0.75 ml ampoules, 99.8 atom% D
Pyridine-d5, for NMR, packaged in 0.75 ml ampoules, 99.8 atom% D
Pyridine-d5, for NMR, packaged in 0.75 ml ampoules, 99.8 atom% D
Thermo Scientific Chemicals
Pyridine-d5, for NMR, packaged in 0.75 ml ampoules, 99.8 atom% D
CAS: 7291-22-7 | C5H5N | 84.13 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA3514800757.5 mLAmpule
Catalog number FSA351480075
Price (MYR)
959.00
Add to cart
Quantity:
7.5 mL
Packaging:
Ampule
Request bulk or custom format
Price (MYR)
959.00
Add to cart
Chemical Identifiers
CAS857730-21-3
IUPAC Name5-bromo-2-chloropyridin-4-amine
Molecular FormulaC5H4BrClN2
InChI KeyMGZWZNBANVSZLM-UHFFFAOYSA-N
SMILESNC1=CC(Cl)=NC=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)122.0-134.0?C
FormPowder
Appearance (Color)White to pale cream
Assay (GC)≥94.0%
For NMR
RUO – Research Use Only