Acetonitrile-d3, for NMR, 99.9 atom% D
Acetonitrile-d3, for NMR, 99.9 atom% D
Acetonitrile-d3, for NMR, 99.9 atom% D
Acetonitrile-d3, for NMR, 99.9 atom% D
Thermo Scientific Chemicals
Acetonitrile-d3, for NMR, 99.9 atom% D
CAS: 2206-26-0 | C2H3N | 44.07 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA35139010010 mLGlass Bottle
Catalog number FSA351390100
Price (MYR)
763.00
Add to cart
Quantity:
10 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
763.00
Add to cart
Specifications
Chemical Name or MaterialAcetonitrile-d3
Density0.8440g/mL
Deuterium (D)99.85% min.
Flash Point2°C
PackagingGlass Bottle
View more
For NMR
RUO – Research Use Only