Cumyl hydroperoxide, 80%, Thermo Scientific Chemicals
Cumyl hydroperoxide, 80%, Thermo Scientific Chemicals
Cumyl hydroperoxide, 80%, Thermo Scientific Chemicals
Cumyl hydroperoxide, 80%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cumyl hydroperoxide, 80%, Thermo Scientific Chemicals
CAS: 80-15-9 | C9H12O2 | 152.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
250 g
1 kg
Packaging:
Plastic bottle
Catalog number FSA349960050
Price (MYR)
283.00
Add to cart
Quantity:
5 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
283.00
Add to cart
Chemical Identifiers
CAS98-83-9
IUPAC Name(prop-1-en-2-yl)benzene
Molecular FormulaC9H10
InChI KeyXYLMUPLGERFSHI-UHFFFAOYSA-N
SMILESCC(=C)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to almost colorless
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.5360 to 1.5400 (20°C, 589 nm)
Stabilizer10 to 20 ppm tert-Butylcatechol
View more
Oxidation reagent
RUO – Research Use Only