Cumyl hydroperoxide, 80%, Thermo Scientific Chemicals
Cumyl hydroperoxide, 80%, Thermo Scientific Chemicals
Cumyl hydroperoxide, 80%, Thermo Scientific Chemicals
Cumyl hydroperoxide, 80%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cumyl hydroperoxide, 80%, Thermo Scientific Chemicals
CAS: 80-15-9 | C9H12O2 | 152.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 g
250 g
Packaging:
Plastic bottle
Catalog number FSA349960010
Price (MYR)
951.00
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
951.00
Add to cart
Chemical Identifiers
CAS2411-89-4
IUPAC Name2-[({5-[1-(3-{[bis(carboxymethyl)amino]methyl}-4-hydroxy-5-methylphenyl)-3-oxo-1,3-dihydro-2-benzofuran-1-yl]-2-hydroxy-3-methylphenyl}methyl)(carboxymethyl)amino]acetic acid
Molecular FormulaC32H32N2O12
InChI KeyIYZPEGVSBUNMBE-UHFFFAOYSA-N
SMILESCC1=CC(=CC(CN(CC(O)=O)CC(O)=O)=C1O)C1(OC(=O)C2=CC=CC=C12)C1=CC(C)=C(O)C(CN(CC(O)=O)CC(O)=O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormPowder
Loss on Drying<6.0% (105?C/constant weight)
lambda-max (UV-Vis)561-581 nm (0.1M NaOH)
Oxidation reagent
RUO – Research Use Only