Celite™ 545, Thermo Scientific Chemicals
Celite™ 545, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Celite™ 545, Thermo Scientific Chemicals
CAS: 68855-54-9 | CNa2O3 | 105.988 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 kg
500 g
2.5 kg
Packaging:
View all
Fiber Drum
Catalog number FSA349670100
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 kg
Packaging:
Fiber Drum
Request bulk or custom format
Chemical Identifiers
CAS7440-44-0
IUPAC Namecarbon
Molecular FormulaC
InChI KeyOKTJSMMVPCPJKN-UHFFFAOYSA-N
SMILES[C]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Black
Appearance (Form)Powder
Loss on drying=<15 %
pH5.5 to 8
Phosphate (PO4)=<3.5 % (by acid extraction)
View more
Filter aid
RUO – Research Use Only