Celite™ 545, Thermo Scientific Chemicals
Celite™ 545, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Celite™ 545, Thermo Scientific Chemicals
CAS: 68855-54-9 | CNa2O3 | 105.988 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
500 g
10 kg
Packaging:
View all
Plastic Drum
Catalog number FSA349670025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 kg
Packaging:
Plastic Drum
Request bulk or custom format
Chemical Identifiers
CAS7439-96-5
IUPAC Namemanganese
Molecular FormulaMn
InChI KeyPWHULOQIROXLJO-UHFFFAOYSA-N
SMILES[Mn]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.05% max.
Filter aid
RUO – Research Use Only