1H,1H,2H,2H-Heptadecafluorodecyl acrylate, 97%, stabilized
1H,1H,2H,2H-Heptadecafluorodecyl acrylate, 97%, stabilized
1H,1H,2H,2H-Heptadecafluorodecyl acrylate, 97%, stabilized
1H,1H,2H,2H-Heptadecafluorodecyl acrylate, 97%, stabilized
Thermo Scientific Chemicals
1H,1H,2H,2H-Heptadecafluorodecyl acrylate, 97%, stabilized
CAS: 27905-45-9 | C13H7F17O2 | 518.17 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA3486100505 gGlass Bottle
Catalog number FSA348610050
Price (MYR)
596.00
Add to cart
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
596.00
Add to cart
Chemical Identifiers
CAS153624-38-5
IUPAC Name[4-(2-methylpropyl)phenyl]boronic acid
Molecular FormulaC10H15BO2
InChI KeyYZSPHVWALUJQNK-UHFFFAOYSA-N
SMILESCC(C)CC1=CC=C(C=C1)B(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%