Lauryl methacrylate, 97%, stabilized, Thermo Scientific Chemicals
Lauryl methacrylate, 97%, stabilized, Thermo Scientific Chemicals
Lauryl methacrylate, 97%, stabilized, Thermo Scientific Chemicals
Lauryl methacrylate, 97%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Lauryl methacrylate, 97%, stabilized, Thermo Scientific Chemicals
CAS: 142-90-5 | C16H30O2 | 254.41 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
Packaging:
Glass bottle
Catalog number FSA347715000
Price (MYR)
847.00
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
847.00
Add to cart
Chemical Identifiers
CAS409-21-2
IUPAC Namemethanidylidynesilylium
Molecular FormulaCSi
InChI KeyHBMJWWWQQXIZIP-UHFFFAOYSA-N
SMILES[C-]#[Si+]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥98.0% SiC
Impurity contentIron(III) oxide Fe2O3: ≤0.25%
Particle SizeMean (d50) 29.2 ? 1.5?m
Appearance (Color)Dark grey to black
Particle SizeConforms to FEPA-Standard 42-1:2006