Brij™ 98, Thermo Scientific Chemicals
Brij™ 98, Thermo Scientific Chemicals
Brij™ 98, Thermo Scientific Chemicals
Brij™ 98, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Brij™ 98, Thermo Scientific Chemicals
CAS: 9004-98-2 | C58H116O21 | 1149.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass bottle
Catalog number FSA347185000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialBrij™ 98
ColorWhite to Cream
Acid Value1mg KOH/g max.
CAS9004-98-2
Physical FormSoft Waxy Solid