all-trans-Retinyl acetate, 1.5 million IU/g, in sunflower oil, stabilized
all-trans-Retinyl acetate, 1.5 million IU/g, in sunflower oil, stabilized
all-trans-Retinyl acetate, 1.5 million IU/g, in sunflower oil, stabilized
all-trans-Retinyl acetate, 1.5 million IU/g, in sunflower oil, stabilized
Thermo Scientific Chemicals
all-trans-Retinyl acetate, 1.5 million IU/g, in sunflower oil, stabilized
CAS: 127-47-9 | C22H32O2 | 328.49 g/mol
Have Questions?
Catalog NumberQuantity
FSA346321000100 g
Catalog number FSA346321000
Price (MYR)
1,092.00
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
1,092.00
Add to cart
Chemical Identifiers
CAS94108-56-2
IUPAC Name4-(trifluoromethoxy)benzene-1-sulfonyl chloride
Molecular FormulaC7H4ClF3O3S
InChI KeyUHCDBMIOLNKDHG-UHFFFAOYSA-N
SMILESFC(F)(F)OC1=CC=C(C=C1)S(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.4775-1.4815 @ 20?C
Assay (GC)≥97.5%
Appearance (Color)Clear colorless to pale yellow