4-(tert-Butyl)benzylchloride, 97%, Thermo Scientific Chemicals
4-(tert-Butyl)benzylchloride, 97%, Thermo Scientific Chemicals
4-(tert-Butyl)benzylchloride, 97%, Thermo Scientific Chemicals
4-(tert-Butyl)benzylchloride, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-(tert-Butyl)benzylchloride, 97%, Thermo Scientific Chemicals
CAS: 19692-45-6 | C11H15Cl | 182.69 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
View all
Glass Bottle
Catalog number FSA345490250
Price (MYR)
1,209.00
Add to cart
Quantity:
25 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
1,209.00
Add to cart
Specifications
Chemical Name or Material4-(tert-Butyl)benzylchloride
Name Note97%
CAS19692-45-6
CAS Min %96.0
Health Hazard 1GHS Signal Word: Danger