1,2,3-1H-Triazole, 97%, Thermo Scientific Chemicals
1,2,3-1H-Triazole, 97%, Thermo Scientific Chemicals
1,2,3-1H-Triazole, 97%, Thermo Scientific Chemicals
1,2,3-1H-Triazole, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,2,3-1H-Triazole, 97%, Thermo Scientific Chemicals
CAS: 288-36-8 | C2H3N3 | 69.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number FSA345480010
Price (MYR)
278.00
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
278.00
Add to cart
Chemical Identifiers
CAS1109-15-5
IUPAC Nametris(2,3,4,5,6-pentafluorophenyl)borane
Molecular FormulaC18BF15
InChI KeyOBAJXDYVZBHCGT-UHFFFAOYSA-N
SMILESFC1=C(F)C(F)=C(B(C2=C(F)C(F)=C(F)C(F)=C2F)C2=C(F)C(F)=C(F)C(F)=C2F)C(F)=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Infrared spectrumConforms
NMR>=94 %
Appearance (Color)White to beige