Brij™ C20, Thermo Scientific Chemicals
Brij™ C20, Thermo Scientific Chemicals
Brij™ C20, Thermo Scientific Chemicals
Brij™ C20, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Brij™ C20, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
Catalog number FSA344295000
Price (MYR)
493.00
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
493.00
Add to cart
Chemical Identifiers
CAS13826-86-3
IUPAC Namedioxoamine; tetrafluoroboranuide
Molecular FormulaBF4NO2
InChI KeyXYRQURYIJKJVSF-UHFFFAOYSA-N
SMILES[O-][NH+]=O.F[B-](F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to orange
FormLiquid
Assay (unspecified)0.30-0.55 M