Pyridine, 99.5%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Pyridine, 99.5%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Pyridine, 99.5%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Pyridine, 99.5%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Pyridine, 99.5%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Pyridine, 99.5%, C5H5N, CAS Number-110-86-1, azabenzene, azine, caswell no. 717, piridina, piridina italian, pirydyna, pirydyna polish, pyridin, pyridin german, rcra waste number u196, 2.5L, 20, 181, 115.0 deg.C to 116.0 deg.C, CHEBI:16227, Colorless, 0.9780g/mL
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
100 mL
1 L
Packaging:
View all
Glass Bottle
Catalog number FSA339420025
Price (MYR)
2,003.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
2,003.00
Add to cart
Specifications
Chemical Name or MaterialPyridine
Melting Point-42.0°C
CAS110-86-1
ColorColorless
Health Hazard 1Danger