n-Hexadecyl palmitate, 98%, Thermo Scientific Chemicals
n-Hexadecyl palmitate, 98%, Thermo Scientific Chemicals
n-Hexadecyl palmitate, 98%, Thermo Scientific Chemicals
n-Hexadecyl palmitate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
n-Hexadecyl palmitate, 98%, Thermo Scientific Chemicals
CAS: 540-10-3 | C32H64O2 | 480.86 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number FSA338461000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS16013-85-7
IUPAC Name2,6-dichloro-3-nitropyridine
Molecular FormulaC5H2Cl2N2O2
InChI KeySHCWQWRTKPNTEM-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=C(Cl)N=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point58°C to 68°C
Appearance (Form)Powder or crystals and/or chunks
GC>=88 %
Appearance (Color)Yellow to orange-brown
Water=<10 % (K.F.)