4-Methylumbelliferyl-β-D-glucuronide dihydrate, 98%, Thermo Scientific Chemicals
4-Methylumbelliferyl-β-D-glucuronide dihydrate, 98%, Thermo Scientific Chemicals
4-Methylumbelliferyl-β-D-glucuronide dihydrate, 98%, Thermo Scientific Chemicals
4-Methylumbelliferyl-β-D-glucuronide dihydrate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Methylumbelliferyl-β-D-glucuronide dihydrate, 98%, Thermo Scientific Chemicals
CAS: 6160-80-1 | C16H16O9·2H2O | 388.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number FSA337220010
Price (MYR)
448.00
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
448.00
Add to cart
Specifications
Chemical Name or Material4-Methylumbelliferyl-β-D-glucuronide dihydrate
CAS6160-80-1
Melting Point102.0°C
EINECS Number228-186-
Infrared SpectrumAuthentic