2-Hydroxyethyl acrylate, 97%, stabilized, Thermo Scientific Chemicals
2-Hydroxyethyl acrylate, 97%, stabilized, Thermo Scientific Chemicals
2-Hydroxyethyl acrylate, 97%, stabilized, Thermo Scientific Chemicals
2-Hydroxyethyl acrylate, 97%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Hydroxyethyl acrylate, 97%, stabilized, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Catalog number FSA334082500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS12054-85-2
IUPAC Nameammonium hydrate molybdenum oxidandiide
Molecular FormulaH6MoNO2
InChI KeyXCOMAVCHADVANQ-UHFFFAOYSA-O
SMILES[NH4+].O.[O--].[Mo]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Titration Complexometric81.0 to 83.0 % (as MoO3)
Insoluble matter=<0.005 %
Nitrate (NO3)(Limit about 0.003%) Passes test
AsO4, PO4, SiO4 (as SiO2)=<0.001 %