Dess-Martin periodinane, 15 wt.% solution in dichloromethane, Thermo Scientific Chemicals
Dess-Martin periodinane, 15 wt.% solution in dichloromethane, Thermo Scientific Chemicals
Dess-Martin periodinane, 15 wt.% solution in dichloromethane, Thermo Scientific Chemicals
Dess-Martin periodinane, 15 wt.% solution in dichloromethane, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Dess-Martin periodinane, 15 wt.% solution in dichloromethane, Thermo Scientific Chemicals
Dess-Martin periodinane, 10 to 15%, C13H13IO8, CAS Number-87413-09-0, 75-09-2, dess martin, dess-martin, dess-martinperiodinane, dess-martin periodinane, 1,1,1-triacetoxy-1,1-dihydro-1,2-benziodoxol-3 1h-one, dess martin periodinane, dess-martin periodane, dess-martin reagent, triacetoxyperiodinane
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mL
Catalog number FSA333110100
Price (MYR)
475.00
Add to cart
Quantity:
10 mL
Request bulk or custom format
Price (MYR)
475.00
Add to cart
Chemical Identifiers
CAS90-01-7
IUPAC Name2-(hydroxymethyl)phenol
Molecular FormulaC7H8O2
InChI KeyCQRYARSYNCAZFO-UHFFFAOYSA-N
SMILESOCC1=CC=CC=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or flakes
Appearance (Color)Light beige
Infrared spectrumConforms
GC>=96.0 %
Melting point83°C to 87°C
oxidation reagent
RUO – Research Use Only