Lithium hydride, 97%, pure, powder (<100 μm)
Lithium hydride, 97%, pure, powder (&lt;100 &mu;m)
Lithium hydride, 97%, pure, powder (&lt;100 &mu;m)
Lithium hydride, 97%, pure, powder (&lt;100 &mu;m)
Lithium hydride, 97%, pure, powder (<100 μm)
CAS: 7580-67-8 | HLi | 7.95 g/mol
Have Questions?
Catalog NumberQuantity
FSA332955000500 g
Catalog number FSA332955000
Price (MYR)
932.35
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
932.35
Add to cart
Chemical Identifiers
CAS360-97-4
IUPAC Name4-amino-1H-imidazole-5-carboxamide
Molecular FormulaC4H6N4O
InChI KeyDVNYTAVYBRSTGK-UHFFFAOYSA-N
SMILESNC(=O)C1=C(N)N=CN1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Grey to purple
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=94 %
Reduction reagent
RUO – Research Use Only