Iodoethane, 98%, pure, stabilized with silver, Thermo Scientific Chemicals
Iodoethane, 98%, pure, stabilized with silver, Thermo Scientific Chemicals
Iodoethane, 98%, pure, stabilized with silver, Thermo Scientific Chemicals
Iodoethane, 98%, pure, stabilized with silver, Thermo Scientific Chemicals
Iodoethane, 98%, pure, stabilized with silver, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Iodoethane, 98%, pure, stabilized with silver, Thermo Scientific Chemicals
CAS: 75-03-6 | C2H5I | 155.97 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass bottle
Catalog number FSA332125000
Price (MYR)
1,202.00
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,202.00
Add to cart
Chemical Identifiers
CAS24057-28-1
IUPAC Name4-methylbenzene-1-sulfonic acid; pyridine
Molecular FormulaC12H13NO3S
InChI KeyZDYVRSLAEXCVBX-UHFFFAOYSA-N
SMILESC1=CC=NC=C1.CC1=CC=C(C=C1)S(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light beige
Melting point112°C to 119°C
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
Titration with NaOH>=97.5 %
Alkylation reagent
RUO – Research Use Only