Iodoethane, 98%, pure, stabilized with silver, Thermo Scientific Chemicals
Iodoethane, 98%, pure, stabilized with silver, Thermo Scientific Chemicals
Iodoethane, 98%, pure, stabilized with silver, Thermo Scientific Chemicals
Iodoethane, 98%, pure, stabilized with silver, Thermo Scientific Chemicals
Iodoethane, 98%, pure, stabilized with silver, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Iodoethane, 98%, pure, stabilized with silver, Thermo Scientific Chemicals
CAS: 75-03-6 | C2H5I | 155.97 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number FSA332121000
Price (MYR)
388.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
388.00
Add to cart
Chemical Identifiers
CAS666-52-4
IUPAC Name(²H₆)propan-2-one
Molecular FormulaC3H6O
InChI KeyCSCPPACGZOOCGX-WFGJKAKNSA-N
SMILES[2H]C([2H])([2H])C(=O)C([2H])([2H])[2H]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
D-Enrichment>=99.85 %
Water=<0.03 % (Coulometric)
Alkylation reagent
RUO – Research Use Only