2-Methoxy-5-methylaniline, 95%, Thermo Scientific Chemicals
2-Methoxy-5-methylaniline, 95%, Thermo Scientific Chemicals
2-Methoxy-5-methylaniline, 95%, Thermo Scientific Chemicals
2-Methoxy-5-methylaniline, 95%, Thermo Scientific Chemicals
2-Methoxy-5-methylaniline, 95%, Thermo Scientific Chemicals
CAS: 120-71-8 | C8H11NO | 137.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA332080050
Price (MYR)
64.96
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
64.96
Add to cart
Chemical Identifiers
CAS112-80-1
IUPAC Name(9E)-octadec-9-enoic acid
Molecular FormulaC18H34O2
InChI KeyZQPPMHVWECSIRJ-MDZDMXLPSA-N
SMILESCCCCCCCC\C=C\CCCCCCCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to pale yellow
Assay (GC)≥98.5%
Impurity content≤0.3% ( sum of Linoleic and Linolenic acids)