1-Iododecane, 98%, stabilized, Thermo Scientific Chemicals
1-Iododecane, 98%, stabilized, Thermo Scientific Chemicals
1-Iododecane, 98%, stabilized, Thermo Scientific Chemicals
1-Iododecane, 98%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Iododecane, 98%, stabilized, Thermo Scientific Chemicals
CAS: 2050-77-3 | C10H21I | 268.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
View all
Glass Bottle
Catalog number FSA330365000
Price (MYR)
2,782.00
Add to cart
Quantity:
500 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
2,782.00
Add to cart
Chemical Identifiers
CAS73781-91-6
IUPAC Namemethyl 6-chloropyridine-3-carboxylate
Molecular FormulaC7H6ClNO2
InChI KeyRMEDXVIWDFLGES-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=C(Cl)N=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream to cream to pale brown
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
Melting Point (clear melt)82.0-89.0?C