1-Iododecane, 98%, stabilized, Thermo Scientific Chemicals
1-Iododecane, 98%, stabilized, Thermo Scientific Chemicals
1-Iododecane, 98%, stabilized, Thermo Scientific Chemicals
1-Iododecane, 98%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Iododecane, 98%, stabilized, Thermo Scientific Chemicals
CAS: 2050-77-3 | C10H21I | 268.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
View all
Glass bottle
Catalog number FSA330361000
Price (MYR)
669.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
669.00
Add to cart
Chemical Identifiers
CAS608-31-1
IUPAC Name2,6-dichloroaniline
Molecular FormulaC6H5Cl2N
InChI KeyJDMFXJULNGEPOI-UHFFFAOYSA-N
SMILESNC1=C(Cl)C=CC=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or chunks or fused solid
Assay (GC)≥97.5%
Melting Point (clear melt)35.0-42.0?C
Appearance (Color)White to cream to pale gray