Methanol-d4, for NMR, contains 0.03 v/v% TMS, 99.8% atom D, Thermo Scientific Chemicals
Methanol-d4, for NMR, contains 0.03 v/v% TMS, 99.8% atom D, Thermo Scientific Chemicals
Methanol-d4, for NMR, contains 0.03 v/v% TMS, 99.8% atom D, Thermo Scientific Chemicals
Methanol-d4, for NMR, contains 0.03 v/v% TMS, 99.8% atom D, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methanol-d4, for NMR, contains 0.03 v/v% TMS, 99.8% atom D, Thermo Scientific Chemicals
Methanol-d4, CH4O, CAS Number-811-98-3, methan-d3-ol-d, 2h4 methanol, methyl-d3 alcohol-d, perdeuteromethanol, methanol-d4, tetradeuteromethanol, trideuterio deuteriooxy methane, methanol-d4, 99.8 atom % d, methyl alcohol-d4, cd3od, 50mL, 01, III, 1187, Colorless, 0.8880g/mL, 99.75% min., 212-378-6
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
5 mL
10 mL
25 mL
Packaging:
Glass Bottle
Catalog number FSA329920500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 mL
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS9003-39-8
Molecular Formula(C6H9NO)n
SMILES*-CC(-*)N1CCCC1=O
MDL NumberMFCD01076626
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
IdentificationPasses test
Water=<5.0 % (K.F.)
HPLC=<10 ppm 1-Vinylpyrrolidin-2-one
Peroxides=<400 ppm (Titanyl sulphate method)
View more
For NMR
RUO – Research Use Only