1-Hexyne, 98%, Thermo Scientific Chemicals
1-Hexyne, 98%, Thermo Scientific Chemicals
1-Hexyne, 98%, Thermo Scientific Chemicals
1-Hexyne, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Hexyne, 98%, Thermo Scientific Chemicals
CAS: 693-02-7 | C6H10 | 82.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
Packaging:
Glass bottle
Catalog number FSA329900250
Price (MYR)
443.00
Add to cart
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
443.00
Add to cart
Chemical Identifiers
CAS585-71-7
IUPAC Name(1-bromoethyl)benzene
Molecular FormulaC8H9Br
InChI KeyCRRUGYDDEMGVDY-UHFFFAOYNA-N
SMILESCC(Br)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
FormLiquid
Identification (FTIR)Conforms
Appearance (Color)Clear colorless to pale yellow to yellow to brown to gray
Refractive Index1.5585-1.5635 @ 20?C