G418 sulfate, for biochemistry
G418 sulfate, for biochemistry
G418 sulfate, for biochemistry
G418 sulfate, for biochemistry
Thermo Scientific Chemicals
G418 sulfate, for biochemistry
G418 sulfate, geneticin, CAS # 108321-42-2, is an antibiotic closely related to Gentamicin B, the common aminoglycoside antibiotic.
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Packaging:
Glass Bottle
Catalog number FSA329400010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialG418 sulfate
TypeG418 Sulfate
Physical FormPowder
CAS108321-42-2
Health Hazard 2GHS H Statement:
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
May damage fertility. May damage the unborn child.
View more
General description
• G418 sulfate (geneticin) is an aminoglycoside antibiotic isolated from Micromonospora rhodorangea
• G418 is a bacteriostatic that prevents polypeptide formation by inhibiting the elongation step of polypeptide synthesis
Applications
• G418 sulfate can be used for selection of genetically engineered cells in both eukaryotes and prokaryotes
• Laboratory grade for applications such as biochemistry
RUO – Research Use Only
General References:
  1. Arshad, H.; Patel, Z.; Mehrabian, M.; Bourkas, M.; Al-Azzawi, Z.; Schmitt-Ulms, G., & Watts, J. C. 2021. The aminoglycoside G418 hinders de novo prion infection in cultured cells. The Journal of biological chemistry, 297(3), 101073.
  2. Ferguson, M. W.; Gerak, C.; Chow, C.; Rastelli, E. J.; Elmore, K. E.; Stahl, F.; Hosseini-Farahabadi, S.; Baradaran-Heravi, A.; Coltart, D. M., & Roberge, M. 2019. The antimalarial drug mefloquine enhances TP53 premature termination codon readthrough by aminoglycoside G418. PloS one, 14(5), e0216423.