(±)-Verapamil hydrochloride, 99+%
(±)-Verapamil hydrochloride, 99+%
(±)-Verapamil hydrochloride, 99+%
(±)-Verapamil hydrochloride, 99+%
Thermo Scientific Chemicals
(±)-Verapamil hydrochloride, 99+%
CAS: 152-11-4 | C27H38N2O4·HCl | 491.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number FSA329330010
Price (MYR)
486.00
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
486.00
Add to cart
Chemical Identifiers
CAS609-06-3
IUPAC Name2,3,4,5-tetrahydroxypentanal
Molecular FormulaC5H10O5
InChI KeyPYMYPHUHKUWMLA-UHFFFAOYNA-N
SMILESOCC(O)C(O)C(O)C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Optical Rotation-18° to -20° (c=10 in water)
Assay from Suppliers CofA≥98.5% (Unspecified)
Water Content (Karl Fischer Titration)≤0.5%
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder