Nifedipine, 98+%, Thermo Scientific Chemicals
Nifedipine, 98+%, Thermo Scientific Chemicals
Nifedipine, 98+%, Thermo Scientific Chemicals
Nifedipine, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Nifedipine, 98+%, Thermo Scientific Chemicals
CAS: 21829-25-4 | C17H18N2O6 | 346.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number FSA329270010
Price (MYR)
373.00
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
373.00
Add to cart
Chemical Identifiers
CAS3682-35-7
IUPAC Nametris(pyridin-2-yl)-1,3,5-triazine
Molecular FormulaC18H12N6
InChI KeyKMVWNDHKTPHDMT-UHFFFAOYSA-N
SMILESC1=CC=C(N=C1)C1=NC(=NC(=N1)C1=CC=CC=N1)C1=CC=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to yellow
Appearance (Form)Powder
Infrared spectrumConforms
Melting point248°C to 252°C
Titration with HClO498.5 to 101.5 %