Glipizide, 98+%
Glipizide, 98+%
Glipizide, 98+%
Thermo Scientific Chemicals
Glipizide, 98+%
CAS: 29094-61-9 | C21H27N5O4S | 445.5 g/mol
Have Questions?
Catalog NumberQuantity
FSA3291100101 g
Catalog number FSA329110010
Price (MYR)
375.39
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
375.39
Add to cart
Chemical Identifiers
CAS3483-12-3
IUPAC Name(2R,3R)-1,4-disulfanylbutane-2,3-diol
Molecular FormulaC4H10O2S2
InChI KeyVHJLVAABSRFDPM-IMJSIDKUSA-N
SMILESO[C@@H](CS)[C@@H](O)CS
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Melting point38°C to 43°C
Infrared spectrumConforms
Appearance (Form)Powder or beads
Titration Iodimetric97.5 to 102.5 %