Leupeptin, 90%, synthetic
Leupeptin, 90%, synthetic
Leupeptin, 90%, synthetic
Leupeptin, 90%, synthetic
Thermo Scientific Chemicals
Leupeptin, 90%, synthetic
CAS: 103476-89-7 | 0·5 H2O4S
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mg
25 mg
Catalog number FSA328350050
Price (MYR)
490.00
Add to cart
Quantity:
5 mg
Request bulk or custom format
Price (MYR)
490.00
Add to cart
Chemical Identifiers
CAS504-29-0
IUPAC Namepyridin-2-amine
Molecular FormulaC5H6N2
InChI KeyICSNLGPSRYBMBD-UHFFFAOYSA-N
SMILESNC1=CC=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
FormCrystalline powder, crystals, flakes or lumps
Melting Point (clear melt)55-60?C
Appearance (Color)White to cream to yellow
Assay (GC)≥98.5%