Genistein, 99%, synthetic, Thermo Scientific Chemicals
Genistein, 99%, synthetic, Thermo Scientific Chemicals
Genistein, 99%, synthetic, Thermo Scientific Chemicals
Genistein, 99%, synthetic, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Genistein, 99%, synthetic, Thermo Scientific Chemicals
CAS: 446-72-0 | C15H10O5 | 270.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mg
100 mg
Packaging:
Glass bottle
Catalog number FSA328270250
Price (MYR)
686.00
Add to cart
Quantity:
25 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
686.00
Add to cart
Chemical Identifiers
CAS50-28-2
IUPAC Name(1S,3aS,3bR,9bS,11aS)-11a-methyl-1H,2H,3H,3aH,3bH,4H,5H,9bH,10H,11H,11aH-cyclopenta[a]phenanthrene-1,7-diol
Molecular FormulaC18H24O2
InChI KeyVOXZDWNPVJITMN-ZBRFXRBCSA-N
SMILESC[C@]12CC[C@H]3[C@@H](CCC4=CC(O)=CC=C34)[C@@H]1CC[C@@H]2O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Assay (Silylated GC)> 98.5%
Water Content (Karl Fischer Titration)<4.0%
Optical Rotation+79.0? ? 3.0? (c=1 in Dioxane)
Appearance (Color)White