Bafilomycin A1, 95%
Bafilomycin A1, 95%
Bafilomycin A1, 95%
Thermo Scientific Chemicals
Bafilomycin A1, 95%
Bafilomycin A1, CAS # 88899-55-2, is an inhibitor of vacuolar ATPases from Streptomyces griseus that has antibacterial, antifungal, and antineoplastic activity. | CAS: 88899-55-2 | C35H58O9 | 622.84 g/mol
Have Questions?
Catalog NumberQuantity
FSA3281200010.1 mg
Catalog number FSA328120001
Price (MYR)
2,295.00
Add to cart
Quantity:
0.1 mg
Request bulk or custom format
Price (MYR)
2,295.00
Add to cart
Chemical Identifiers
CAS1313-60-6
IUPAC Namedisodium dioxidanediide
Molecular FormulaNa2O2
InChI KeyPFUVRDFDKPNGAV-UHFFFAOYSA-N
SMILES[Na+].[Na+].[O-][O-]
View more
SpecificationsSpecification SheetSpecification Sheet
AppearanceMay contain a small quantity of discolored
Phosphate (PO4)=<5 ppm
Titration with KMnO4>=93.0 %
Heavy metals (as Pb)=<20 ppm
Sulfate (SO4)=<10 ppm
View more
General Description
  • Bafilomycin A1 is a specific potent inhibitor of vacuolar ATPases
  • It is a macrolide antibiotic, and shows fungicide activity
Application
  • Bafilomycin A1 has antibacterial, antifungal, and antineoplastic activity
  • It can be immunosuppressive
  • It can prevent maturation of autophagic vacuoles by inhibiting fusion between autophagosomes and lysosomes
  • It is a potent and selective inhibitor of vacuolar-type H+ ATPase (V-ATPase)
RUO – Research Use Only
General References:
  1. Miao, Y.; Li, G.; Zhang, X.; Xu, H.; Abraham, S. N. A TRP Channel Senses Lysosome Neutralization by Pathogens to Trigger Their Expulsion. Cell. 2015, 161 (6), 1306-1319.
  2. Lee, K. I.; Kim, M. J.; Koh, H.; Lee, J. I.; Namkoong, S.; Oh, W. K.; Park, J.The anti-hypertensive drug reserpine induces neuronal cell death through inhibition of autophagic flux. Biochem. Biophys. Res. Commun. 2015, 462 (4), 402-408.