AEBSF hydrochloride, 98%
AEBSF hydrochloride, 98%
AEBSF hydrochloride, 98%
AEBSF hydrochloride, 98%
Thermo Scientific Chemicals
AEBSF hydrochloride, 98%
CAS: 30827-99-7 | C8H10FNO2S·ClH | 239.7 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
50 mg
Packaging:
Glass bottle
Catalog number FSA328110010
Price (MYR)
2,984.00
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,984.00
Add to cart
Chemical Identifiers
CAS130250-54-3
IUPAC Name1-tert-butyl 3-ethyl piperidine-1,3-dicarboxylate
Molecular FormulaC13H23NO4
InChI KeyYCXCRFGBFZTUSU-UHFFFAOYNA-N
SMILESCCOC(=O)C1CCCN(C1)C(=O)OC(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormFused solid
Assay (GC)≥96.0%
Melting Point (clear melt)30.0-39.0?C